Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH8
Entrez Gene ID
101111015
Gene Name
retinol dehydrogenase 8
Gene Aliases
-
Chromosome Location
Chr.5: 12867034 - 12870540 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095717.2 XP_014951203.1 1 - 2 152 95

View Full Product Details