Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTD1
Entrez Gene ID
101105930
Gene Name
cyclin N-terminal domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 20300662 - 20308909 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012946.4 XP_004012995.1 5 - 6 890 85

View Full Product Details