Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCRA
Entrez Gene ID
101110628
Gene Name
pre T cell antigen receptor alpha
Gene Aliases
-
Chromosome Location
Chr.20: 18296100 - 18301227 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018826.4 XP_004018875.1 1 - 2 63 71

View Full Product Details