Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SKOR2
Entrez Gene ID
101110895
Gene Name
SKI family transcriptional corepressor 2
Gene Aliases
-
Chromosome Location
Chr.23: 52301169 - 52341563 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960880.1 XP_027816681.1 5 - 6 2900 98

View Full Product Details