Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EME2
Entrez Gene ID
101111068
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
-
Chromosome Location
Chr.24: 1307443 - 1313565 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961939.1 XP_027817740.1 5 - 6 771 97

View Full Product Details