Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CISD3
Entrez Gene ID
101120095
Gene Name
CDGSH iron sulfur domain 3
Gene Aliases
-
Chromosome Location
Chr.11: 23973991 - 23977086 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012823.3 XP_004012872.1 3 - 4 375 68

View Full Product Details