Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC10
Entrez Gene ID
101122425
Gene Name
homeobox C10
Gene Aliases
-
Chromosome Location
Chr.3: 142064111 - 142069228 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006284.4 XP_004006333.2 1 - 2 867 67

View Full Product Details