Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOHLH1
Entrez Gene ID
105607227
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 1
Gene Aliases
-
Chromosome Location
Chr.3: 3782894 - 3787724 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968316.1 XP_027824117.1 6 - 7 1316 61

View Full Product Details