Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRK
Entrez Gene ID
101106946
Gene Name
fyn related Src family tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.8: 24139136 - 24242055 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011186.4 XP_004011235.2 7 - 8 1880 100

View Full Product Details