Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNGR3
Entrez Gene ID
101113121
Gene Name
synaptogyrin 3
Gene Aliases
-
Chromosome Location
Chr.24: 1502519 - 1506700 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104019.2 XP_014959505.2 2 - 3 436 126

View Full Product Details