Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGIX
Entrez Gene ID
101112793
Gene Name
MAGI family member, X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 58308795 - 58312600 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963427.1 XP_027819228.1 6 - 7 931 87

View Full Product Details