Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBX10
Entrez Gene ID
101111676
Gene Name
T-box transcription factor 10
Gene Aliases
-
Chromosome Location
Chr.21: 47231624 - 47240385 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959665.1 XP_027815466.1 7 - 8 899 66

View Full Product Details