Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP5
Entrez Gene ID
101114099
Gene Name
THAP domain containing 5
Gene Aliases
-
Chromosome Location
Chr.4: 66218644 - 66224582 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007887.3 XP_004007936.2 2 - 3 411 135
XM_012176795.2 XP_012032185.2 3 - 4 667 135
XM_027968617.1 XP_027824418.1 2 - 3 238 135
XM_027968618.1 XP_027824419.1 2 - 3 358 135

View Full Product Details