Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOC3L2
Entrez Gene ID
101115304
Gene Name
exocyst complex component 3 like 2
Gene Aliases
-
Chromosome Location
Chr.14: 56377959 - 56409218 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012190547.3 XP_012045937.2 9 - 10 1995 97
XM_027978471.1 XP_027834272.1 9 - 10 1995 97

View Full Product Details