Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
101119235
Gene Name
tumor protein p63 regulated 1 like
Gene Aliases
-
Chromosome Location
Chr.12: 53253476 - 53257121 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099263.2 XP_014954749.2 4 - 5 701 67

View Full Product Details