Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNIP1
Entrez Gene ID
101114317
Gene Name
Smad nuclear interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 12819615 - 12836349 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004001808.4 XP_004001857.2 3 - 4 992 80

View Full Product Details