Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRFN5
Entrez Gene ID
101103927
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Gene Aliases
-
Chromosome Location
Chr.18: 51043745 - 51060906 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957296.1 XP_027813097.1 2 - 3 2098 94

View Full Product Details