Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMG1
Entrez Gene ID
101108377
Gene Name
EMG1 N1-specific pseudouridine methyltransferase
Gene Aliases
-
Chromosome Location
Chr.3: 224257696 - 224263538 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006919.4 XP_004006968.3 2 - 3 445 104
XM_027967923.1 XP_027823724.1 2 - 3 453 104

View Full Product Details