Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFTAP
Entrez Gene ID
101107893
Gene Name
intraflagellar transport associated protein
Gene Aliases
C15H11orf74
Chromosome Location
Chr.15: 72178301 - 72386407 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016413.3 XP_004016462.1 2 - 3 276 99
XM_012096242.3 XP_011951632.1 2 - 3 300 99
XM_012096243.3 XP_011951633.1 4 - 5 410 99
XM_027979635.1 XP_027835436.1 3 - 4 352 99

View Full Product Details