Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMIM20
Entrez Gene ID
101106940
Gene Name
small integral membrane protein 20
Gene Aliases
C6H4orf52
Chromosome Location
Chr.6: 50811044 - 50823654 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009751.3 XP_004009800.1 2 - 3 268 76

View Full Product Details