Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB10
Entrez Gene ID
101112326
Gene Name
zinc finger and BTB domain containing 10
Gene Aliases
-
Chromosome Location
Chr.9: 61752951 - 61785120 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011759.4 XP_004011808.3 3 - 4 1974 109

View Full Product Details