Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB1B
Entrez Gene ID
101104947
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.21: 46104272 - 46111482 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960031.1 XP_027815832.1 5 - 6 502 77

View Full Product Details