Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEM5
Entrez Gene ID
101119950
Gene Name
thioesterase superfamily member 5
Gene Aliases
-
Chromosome Location
Chr.1: 107533278 - 107542040 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002506.4 XP_004002555.1 5 - 6 885 67

View Full Product Details