Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL36G
Entrez Gene ID
101106514
Gene Name
interleukin 36 gamma
Gene Aliases
-
Chromosome Location
Chr.3: 64021207 - 64028971 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012170103.3 XP_012025493.1 3 - 4 307 65
XM_012170108.3 XP_012025498.1 3 - 4 241 65
XM_027966772.1 XP_027822573.1 3 - 4 307 65

View Full Product Details