Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTHD1
Entrez Gene ID
101108656
Gene Name
death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 61279191 - 61369202 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010053.4 XP_004010102.3 4 - 5 1547 110
XM_027970923.1 XP_027826724.1 3 - 4 576 110

View Full Product Details