Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUCL3
Entrez Gene ID
101106634
Gene Name
mucin like 3
Gene Aliases
DPCR1
Chromosome Location
Chr.20: 29907802 - 29917384 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958560.1 XP_027814361.1 2 - 3 598 64

View Full Product Details