Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF23
Entrez Gene ID
101112475
Gene Name
fibroblast growth factor 23
Gene Aliases
-
Chromosome Location
Chr.3: 226514564 - 226525683 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027967989.1 XP_027823790.1 1 - 2 236 74

View Full Product Details