Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPA
Entrez Gene ID
101103949
Gene Name
XPA, DNA damage recognition and repair factor
Gene Aliases
-
Chromosome Location
Chr.2: 52625663 - 52655617 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004232.4 XP_004004281.1 2 - 3 296 140
XM_012120695.3 XP_011976085.1 2 - 3 286 140

View Full Product Details