Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCNN1D
Entrez Gene ID
101105267
Gene Name
sodium channel epithelial 1 subunit delta
Gene Aliases
-
Chromosome Location
Chr.12: 54998900 - 55003086 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976120.1 XP_027831921.1 6 - 7 1232 81

View Full Product Details