Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK1C
Entrez Gene ID
101111509
Gene Name
divergent protein kinase domain 1C
Gene Aliases
FAM69C
Chromosome Location
Chr.23: 4284370 - 4301806 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960694.1 XP_027816495.1 3 - 4 1045 60

View Full Product Details