Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTD
Entrez Gene ID
101113196
Gene Name
biotinidase
Gene Aliases
-
Chromosome Location
Chr.1: 296236178 - 296295009 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961923.1 XP_027817724.1 2 - 3 351 74

View Full Product Details