Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCN3
Entrez Gene ID
101113473
Gene Name
ficolin 3
Gene Aliases
-
Chromosome Location
Chr.2: 255418995 - 255425374 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964876.1 XP_027820677.1 2 - 3 1673 107

View Full Product Details