Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM44
Entrez Gene ID
101114642
Gene Name
tripartite motif containing 44
Gene Aliases
-
Chromosome Location
Chr.15: 71340662 - 71456781 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979631.1 XP_027835432.1 2 - 3 1045 88

View Full Product Details