Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD6
Entrez Gene ID
101116263
Gene Name
tudor domain containing 6
Gene Aliases
-
Chromosome Location
Chr.20: 21581255 - 21592651 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958745.1 XP_027814546.1 1 - 2 6056 79
XM_027958746.1 XP_027814547.1 1 - 2 6056 79

View Full Product Details