Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK9
Entrez Gene ID
101114298
Gene Name
kallikrein related peptidase 9
Gene Aliases
-
Chromosome Location
Chr.14: 60482945 - 60490732 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012094666.3 XP_011950056.2 2 - 3 1606 79

View Full Product Details