Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5DC4
Entrez Gene ID
101106763
Gene Name
5'-nucleotidase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.3: 64269765 - 64279977 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965894.1 XP_027821695.1 2 - 3 243 67

View Full Product Details