Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA9
Entrez Gene ID
101109613
Gene Name
spermatogenesis associated 9
Gene Aliases
-
Chromosome Location
Chr.5: 100868659 - 100924841 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096204.2 XP_014951690.2 1 - 2 62 118

View Full Product Details