Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101104728
Entrez Gene ID
101104728
Gene Name
antimicrobial peptide NK-lysin
Gene Aliases
-
Chromosome Location
Chr.3: 61562380 - 61566125 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965892.1 XP_027821693.1 3 - 4 257 95

View Full Product Details