Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC51A
Entrez Gene ID
100009015
Gene Name
solute carrier family 51 subunit alpha
Gene Aliases
OSTALPA
Chromosome Location
Chr.14: 92519955 - 92534122 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002716536.3 XP_002716582.1 7 - 8 1123 63
GenBank mRNA DQ122755.1 - 4 - 5 444 63

View Full Product Details