Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITGA2B
Entrez Gene ID
100008734
Gene Name
integrin subunit alpha 2b
Gene Aliases
GPIIB
Chromosome Location
Chr.19: 44648352 - 44660521 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082065.1 NP_001075534.1 20 - 21 2124 137
GenBank mRNA AF170528.1 - 20 - 21 2124 137

View Full Product Details