Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGA2B
Entrez Gene ID
100008734
Gene Name
integrin subunit alpha 2b
Gene Aliases
GPIIB
Chromosome Location
Chr.19: 44648352 - 44660521 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082065.1 NP_001075534.1 8 - 9 870 78
GenBank mRNA AF170528.1 - 8 - 9 870 78

View Full Product Details