Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP4B1
Entrez Gene ID
100008805
Gene Name
CYP4B1-like isozyme short form
Gene Aliases
-
Chromosome Location
Chr.13: 119759447 - 119779224 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082103.1 NP_001075572.1 2 - 3 337 74
GenBank mRNA AF176914.5 - 2 - 3 337 74
AF332576.4 - 2 - 3 337 74
M29852.1 - 2 - 3 328 74

View Full Product Details