Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GC
Entrez Gene ID
100008994
Gene Name
GC vitamin D binding protein
Gene Aliases
DBP
Chromosome Location
Chr.15: 78245193 - 78292809 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082204.1 NP_001075673.1 4 - 5 486 83
GenBank mRNA D29666.1 - 4 - 5 486 83

View Full Product Details