Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLMAP
Entrez Gene ID
100009199
Gene Name
sarcolemma associated protein
Gene Aliases
SLAP
Chromosome Location
Chr.9: 24582901 - 24751719 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082348.2 NP_001075817.2 8 - 9 828 125
XM_008260454.2 XP_008258676.1 9 - 10 2064 125
XM_008260455.2 XP_008258677.1 9 - 10 2058 125
XM_008260456.2 XP_008258678.1 9 - 10 2064 125
XM_008260457.2 XP_008258679.1 9 - 10 2064 125
XM_008260458.2 XP_008258680.1 9 - 10 2064 125
XM_008260459.2 XP_008258681.1 9 - 10 2058 125
XM_008260460.2 XP_008258682.1 9 - 10 2064 125
XM_008260461.2 XP_008258683.1 9 - 10 2058 125
XM_008260462.2 XP_008258684.1 9 - 10 2058 125
XM_008260463.2 XP_008258685.1 9 - 10 2064 125
XM_008260464.2 XP_008258686.1 9 - 10 2064 125
XM_008260465.2 XP_008258687.1 9 - 10 2064 125
XM_008260466.2 XP_008258688.1 9 - 10 2066 125
XM_008260467.2 XP_008258689.1 9 - 10 2063 125
XM_008260468.2 XP_008258690.1 9 - 10 2057 125
XM_008260470.2 XP_008258692.1 9 - 10 2066 125
XM_008260471.2 XP_008258693.1 9 - 10 2059 125
XM_008260472.2 XP_008258694.1 9 - 10 2065 125
XM_008260473.2 XP_008258695.1 9 - 10 2059 125
XM_017343638.1 XP_017199127.1 9 - 10 2058 125
XM_017343639.1 XP_017199128.1 9 - 10 2069 125
GenBank mRNA U21157.1 - 8 - 9 1189 125

View Full Product Details