Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC12A4
Entrez Gene ID
100009441
Gene Name
solute carrier family 12 member 4
Gene Aliases
RBKCC1
Chromosome Location
Chr.5: 23506663 - 23528027 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082703.2 NP_001076172.1 15 - 16 1987 61
XM_008257419.1 XP_008255641.1 16 - 17 2060 61
XM_017342181.1 XP_017197670.1 11 - 12 2247 61
GenBank mRNA U55053.1 - 15 - 16 1988 61

View Full Product Details