Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK5
Entrez Gene ID
100009446
Gene Name
potassium two pore domain channel subfamily K member 5
Gene Aliases
-
Chromosome Location
Chr.12: 29027158 - 29065702 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082705.2 NP_001076174.1 2 - 3 407 89
XM_017344780.1 XP_017200269.1 3 - 4 378 89
GenBank mRNA AY752983.1 - 2 - 3 407 89

View Full Product Details