Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC5
Entrez Gene ID
100009574
Gene Name
transient receptor potential cation channel subfamily C member 5
Gene Aliases
TRP5
Chromosome Location
Chr.X: 60237397 - 60560540 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082779.1 NP_001076248.1 7 - 8 2673 100
GenBank mRNA AJ006203.1 - 7 - 8 2673 100

View Full Product Details