Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A7
Entrez Gene ID
100009335
Gene Name
solute carrier family 22 member 7
Gene Aliases
OAT2
Chromosome Location
Chr.12: 32747223 - 32752846 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082642.1 NP_001076111.1 8 - 9 1377 63
XM_017344777.1 XP_017200266.1 8 - 9 1445 63
XM_017344778.1 XP_017200267.1 9 - 10 1180 63
GenBank mRNA DQ146940.1 - 8 - 9 1377 63

View Full Product Details