Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIO1
Entrez Gene ID
100101570
Gene Name
iodothyronine deiodinase 1
Gene Aliases
-
Chromosome Location
Chr.13: 112874983 - 112888996 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099958.1 NP_001093428.1 2 - 3 532 150
GenBank mRNA EF428025.1 - 2 - 3 532 150

View Full Product Details