Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBA2
Entrez Gene ID
100101619
Gene Name
crystallin beta A2
Gene Aliases
-
Chromosome Location
Chr.7: 160231716 - 160234453 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099970.1 NP_001093440.1 3 - 4 340 124
GenBank mRNA EF457886.1 - 3 - 4 340 124

View Full Product Details